Gene Transcripts

Gene Symbol
Rnf19a
Entrez Gene ID
30945
Gene Name
ring finger protein 19A
Gene Aliases
AA032313, Dorfin, Rnf19, UIP117, Ubce7ip2, XYbp
Chromosome Location
Chr. 15: 36239933 - 36283723 on Build GRCm38
UniGene ID
Mm.5181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013923.2 NP_038951.1 8 1888
GenBank mRNA AF120206.1 - 1781
AF120207.1 - 7 1781
AK147768.1 - 8 1876
AK190444.1 - 2 128
BC040769.1 - 7 1663 4457807
DV656092.1 - 3 372
X71642.1 - 4 435

View Full Product Details