Gene Transcripts

Gene Symbol
Aim2
Entrez Gene ID
383619
Gene Name
absent in melanoma 2
Gene Aliases
Gm1313, Ifi210
Chromosome Location
Chr. 1: 173349246 - 173466040 on Build GRCm38
UniGene ID
Mm.131453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013779.2 NP_001013801.2 6 816
GenBank mRNA DT921984.1 - 2 342

View Full Product Details