Gene Transcripts

Gene Symbol
Cul9
Entrez Gene ID
78309
Gene Name
cullin 9
Gene Aliases
1810035I07Rik, Cul-9, Parc, mKIAA0708
Chromosome Location
Chr. 17: 46500609 - 46546388 on Build GRCm38
UniGene ID
Mm.329076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081335.2 NP_001074804.2 4 962
GenBank mRNA AK147675.1 - 4 1410

View Full Product Details