Gene Transcripts

Gene Symbol
Tslp
Entrez Gene ID
53603
Gene Name
thymic stromal lymphopoietin
Gene Aliases
Chromosome Location
Chr. 18: 32815383 - 32819799 on Build GRCm38
UniGene ID
Mm.143716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021367.2 NP_067342.1 2, 3 170
GenBank mRNA AF232937.1 - 2, 3 170
BC130230.1 - 2, 3 174 40141922

View Full Product Details