Gene Transcripts

Gene Symbol
Stmn4
Entrez Gene ID
56471
Gene Name
stathmin-like 4
Gene Aliases
Lagl, R75359, RB3
Chromosome Location
Chr. 14: 66344303 - 66361680 on Build GRCm38
UniGene ID
Mm.35474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019675.2 NP_062649.1 5 563
GenBank mRNA AF105222.1 - 4 535
BC014707.1 - 5 565 4507015
BC046752.1 - 5 563 5693069

View Full Product Details