Gene Transcripts

Gene Symbol
Ubxn4
Entrez Gene ID
67812
Gene Name
UBX domain protein 4
Gene Aliases
1300013G12Rik, Ubxd2, mKIAA0242
Chromosome Location
Chr. 1: 128243898 - 128279378 on Build GRCm38
UniGene ID
Mm.293321
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026390.2 NP_080666.2 6, 7 884
GenBank mRNA AK011233.1 - 6, 7 781
AK019371.1 - 6, 7 777
AK029431.1 - 6, 7 930
AK032206.1 - 6, 7 763
AK075716.1 - 6, 7 619
AK145551.1 - 3, 4 310
AK148333.1 - 770
AK207698.1 - 3, 4 313
BC012271.1 - 6, 7 896
BC019795.1 - 6, 7 903 4158848
BC030190.1 - 893

View Full Product Details