Gene Transcripts

Gene Symbol
Cgrrf1
Entrez Gene ID
68755
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
1110038G02Rik, 1810009H17Rik, CGR19
Chromosome Location
Chr. 14: 46832093 - 46854200 on Build GRCm38
UniGene ID
Mm.343809
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026832.3 NP_081108.2 6 966
GenBank mRNA AK030755.1 - 6 968
AK154797.1 - 6 1042
BC037677.1 - 6 982 3491666

View Full Product Details