Gene Transcripts
- Gene Symbol
- Naa40
- Entrez Gene ID
- 70999
- Gene Name
- N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae)
- Gene Aliases
- 4931433E08Rik, AU023197, Nat11
- Chromosome Location
- Chr. 19: 7225668 - 7241222 on Build GRCm38
- UniGene ID
- Mm.329605
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027643.1 |
NP_081919.1
|
3, 4 |
436 |
|
GenBank mRNA |
AK016504.1 |
- |
3, 4 |
397 |
|
AK033510.1 |
- |
3, 4 |
258 |
|
AK033896.1 |
- |
3, 4 |
267 |
|
AK035418.1 |
- |
2, 3 |
1154 |
|
AK162625.1 |
- |
3, 4 |
270 |
|
AK170548.1 |
- |
3, 4 |
262 |
|
AK213481.1 |
- |
2, 3 |
163 |
|
AK216197.1 |
- |
2, 3 |
164 |
|
BC020020.1 |
- |
3, 4 |
287 |
3156924 |
View Full Product Details