Gene Transcripts

Gene Symbol
Ephx3
Entrez Gene ID
71932
Gene Name
epoxide hydrolase 3
Gene Aliases
2310063B19Rik, Abhd9
Chromosome Location
Chr. 17: 32183770 - 32189850 on Build GRCm38
UniGene ID
Mm.176240
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033163.3 NP_001028335.1 5 864
GenBank mRNA AK132483.1 - 5 865
BC145797.1 - 5 929 40131100

View Full Product Details