Gene Transcripts

Gene Symbol
Ehhadh
Entrez Gene ID
74147
Gene Name
enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase
Gene Aliases
1300002P22Rik, HD, L-PBE, LBFP, LBP, MFP, MFP1, PBFE
Chromosome Location
Chr. 16: 21761285 - 21787834 on Build GRCm38
UniGene ID
Mm.28100
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023737.3 NP_076226.2 6 772
GenBank mRNA AK004867.1 - 6 743
BC016899.1 - 6 746 4241677

View Full Product Details