Gene Transcripts

Gene Symbol
Iqce
Entrez Gene ID
74239
Gene Name
IQ motif containing E
Gene Aliases
1700028P05Rik, mKIAA1023
Chromosome Location
Chr. 5: 140662553 - 140702436 on Build GRCm38
UniGene ID
Mm.179704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028833.3 NP_083109.2 21 2118
GenBank mRNA AK006472.2 - 20 1859
BC059223.1 - 21 2118 6417270
BC065128.1 - 17 4683

View Full Product Details