Gene Transcripts

Gene Symbol
Efcab11
Entrez Gene ID
78767
Gene Name
EF-hand calcium binding domain 11
Gene Aliases
2610021K21Rik, 2610110A13Rik
Chromosome Location
Chr. 12: 99692752 - 99884445 on Build GRCm38
UniGene ID
Mm.235920
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030172.2 NP_084448.1 4 361
GenBank mRNA AK011507.1 - 4 361
AK011836.1 - 4 343
AK035051.1 - 4 341
BC049109.1 - 4 323 6414718

View Full Product Details