Gene Transcripts

Gene Symbol
Gsdmc
Entrez Gene ID
83492
Gene Name
gasdermin C
Gene Aliases
Gsdmc1, Mlze
Chromosome Location
Chr. 15: 63775971 - 63808739 on Build GRCm38
UniGene ID
Mm.335619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031378.3 NP_113555.1 2 459
GenBank mRNA AB042406.1 - 1 356
AK137662.1 - 2 460

View Full Product Details