Gene Transcripts

Gene Symbol
Thtpa
Entrez Gene ID
105663
Gene Name
thiamine triphosphatase
Gene Aliases
AW122966
Chromosome Location
Chr. 14: 55094784 - 55098995 on Build GRCm38
UniGene ID
Mm.319204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153083.5 NP_694723.1 1 868
GenBank mRNA AF432864.2 - 1 699
AK085173.1 - 1 563
AK132479.1 - 1 560
BC025562.1 - 1 852 5322971

View Full Product Details