Gene Transcripts
- Gene Symbol
- Adrbk1
- Entrez Gene ID
- 110355
- Gene Name
- adrenergic receptor kinase, beta 1
- Gene Aliases
- Adrbk-1, Bark-1, GRK2, beta ARK, betaARK1
- Chromosome Location
- Chr. 19: 4285999 - 4306222 on Build GRCm38
- UniGene ID
- Mm.254144
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290818.1 |
NP_001277747.1
|
20, 21 |
2102 |
|
NM_130863.2 |
NP_570933.1
|
21, 22 |
2174 |
|
GenBank mRNA |
AF333028.1 |
- |
20, 21 |
1894 |
|
AK153543.1 |
- |
2, 3 |
126 |
|
AK155691.1 |
- |
20, 21 |
2103 |
|
AK183679.1 |
- |
1, 2 |
105 |
|
BC003196.1 |
- |
20, 21 |
1890 |
|
BC033272.1 |
- |
17, 18 |
2250 |
|
BC038312.1 |
- |
2, 3 |
773 |
|
BC053922.1 |
- |
21, 22 |
1907 |
6309555 |
View Full Product Details