Gene Transcripts

Gene Symbol
Klf1
Entrez Gene ID
16596
Gene Name
Kruppel-like factor 1 (erythroid)
Gene Aliases
Eklf, Nan
Chromosome Location
Chr. 8: 84901928 - 84905295 on Build GRCm38
UniGene ID
Mm.4847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK158037.1 - 2 221
BC114978.1 - 2 260 40046272
M97200.1 - 2 260

View Full Product Details