Gene Transcripts

Gene Symbol
Lat
Entrez Gene ID
16797
Gene Name
linker for activation of T cells
Gene Aliases
Chromosome Location
Chr. 7: 126363827 - 126369589 on Build GRCm38
UniGene ID
Mm.10280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010689.2 NP_034819.1 11 991
GenBank mRNA AF036907.1 - 11 991
BC013337.1 - 11 968 4191983

View Full Product Details