Gene Transcripts

Gene Symbol
Rhox5
Entrez Gene ID
18617
Gene Name
reproductive homeobox 5
Gene Aliases
AA409564, Pem
Chromosome Location
Chr. X: 37754608 - 37808878 on Build GRCm38
UniGene ID
Mm.423066
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008818.2 NP_032844.2 2 513
GenBank mRNA AK146379.1 - 2 538
AK146393.1 - 2 537
AK146449.1 - 2 519
BC056204.1 - 3 521 30022528
DQ058642.1 - 2 497
M32484.1 - 2 497

View Full Product Details