Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004150.3 | NP_001004150.1 | 3 | 1124 | |
NM_001170954.1 | NP_001164425.1 | 3 | 1164 | ||
GenBank mRNA | AK139833.1 | - | 3 | 1128 | |
AK149186.1 | - | 3 | 1080 | ||
AK162529.1 | - | 3 | 1163 | ||
AY371179.1 | - | 3 | 1083 | ||
BC138844.1 | - | 2 | 1039 | 8861866 | |
BC138845.1 | - | 2 | 1039 | 8861867 | |
DQ143885.1 | - | 1 | 858 |