Gene Transcripts
- Gene Symbol
- Anks1b
- Entrez Gene ID
- 77531
- Gene Name
- ankyrin repeat and sterile alpha motif domain containing 1B
- Gene Aliases
- AIDA-1b, C030032C09Rik, E530015N03Rik, ENSMUSG00000078431, Gm10937, Gm1555
- Chromosome Location
- Chr. 10: 89873509 - 90973300 on Build GRCm38
- UniGene ID
- Mm.440605
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036288.1 |
- |
3 |
2426 |
|
View Full Product Details