Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025954.3 | NP_080230.2 | 2 | 989 | |
GenBank mRNA | AK005906.1 | - | 2 | 661 | |
AK010094.1 | - | 2 | 386 | ||
AK012010.1 | - | 213 | |||
AK013578.1 | - | 752 | |||
BC040100.1 | - | 2 | 989 | 4456284 | |
BC083113.1 | - | 1 | 1119 | 5356187 |