Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164640.1 | NP_001158112.1 | 5 | 510 | |
NM_029419.2 | NP_083695.2 | 6 | 610 | ||
GenBank mRNA | AK018646.1 | - | 6 | 612 | |
AK085956.1 | - | 5 | 487 | ||
BC139137.1 | - | 5 | 389 | 8862159 | |
BC139138.1 | - | 5 | 389 | 8862160 |