Gene Transcripts

Gene Symbol
Tra2a
Entrez Gene ID
101214
Gene Name
transformer 2 alpha homolog (Drosophila)
Gene Aliases
1500010G04Rik, AL022798, G430041M01Rik, mAWMS1
Chromosome Location
Chr. 6: 49243921 - 49264248 on Build GRCm38
UniGene ID
Mm.196598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198102.2 NP_932770.2 4 575
GenBank mRNA AK089980.1 - 4 554
AK171708.1 - 4 557
BC057448.1 - 4 533 4016620
BC058764.1 - 5 840

View Full Product Details