Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139142.2 | NP_631881.1 | 11 | 1763 | |
GenBank mRNA | AJ428067.1 | - | 1776 | ||
AJ964961.1 | - | 11 | 1737 | ||
AK043297.1 | - | 11 | 1774 | ||
AK080979.1 | - | 1776 | |||
AK140445.1 | - | 1774 | |||
AK160468.1 | - | 1774 | |||
BC118933.2 | - | 11 | 1763 | 40090800 |