Gene Transcripts
- Gene Symbol
- Ppif
- Entrez Gene ID
- 105675
- Gene Name
- peptidylprolyl isomerase F (cyclophilin F)
- Gene Aliases
- AW457192, CyP-D, CyP-F, CypD, PPIase
- Chromosome Location
- Chr. 14: 25693574 - 25700468 on Build GRCm38
- UniGene ID
- Mm.41656
- Species
- Mus musculus
- Gene Symbol
- Ppifos
- Entrez Gene ID
- 68173
- Gene Name
- peptidylprolyl isomerase F, opposite strand
- Gene Aliases
- 4930520F12Rik, Ppifs
- Chromosome Location
- Chr. 14: 25696356 - 25701282 on Build GRCm38
- UniGene ID
- Mm.444727
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015792.1 |
- |
3 |
483 |
|
AK019686.1 |
- |
3 |
483 |
|
View Full Product Details