Gene Transcripts
- Gene Symbol
- Cdca2
- Entrez Gene ID
- 108912
- Gene Name
- cell division cycle associated 2
- Gene Aliases
- 2610311M19Rik, AI586158
- Chromosome Location
- Chr. 14: 67673180 - 67715841 on Build GRCm38
- UniGene ID
- Mm.33831
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001110162.1 |
NP_001103632.1
|
7, 8 |
1180 |
|
NM_175384.4 |
NP_780593.3
|
7, 8 |
883 |
|
GenBank mRNA |
AK031083.1 |
- |
7, 8 |
946 |
|
AK031313.1 |
- |
3, 4 |
325 |
|
AK160223.1 |
- |
7, 8 |
942 |
|
AK166952.1 |
- |
7, 8 |
949 |
|
BC037136.1 |
- |
7, 8 |
980 |
|
BC067009.1 |
- |
7, 8 |
911 |
|
BC075682.1 |
- |
7, 8 |
883 |
|
BC116298.1 |
- |
3, 4 |
325 |
40095010 |
BC116299.1 |
- |
3, 4 |
325 |
40095011 |
View Full Product Details