Gene Transcripts

Gene Symbol
Ms4a7
Entrez Gene ID
109225
Gene Name
membrane-spanning 4-domains, subfamily A, member 7
Gene Aliases
9130422I10Rik, A430103C15Rik, CD20l4, CFFMA
Chromosome Location
Chr. 19: 11321039 - 11336146 on Build GRCm38
UniGene ID
Mm.491251
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025610.4 NP_001020781.1 6 768
NM_001276398.1 NP_001263327.1 7 1065
NM_027836.6 NP_082112.3 7 825
GenBank mRNA AK018686.1 - 6 739
AK046559.1 - 5 3563
AK079888.1 - 7 1066
AK149890.1 - 7 793
AK151261.1 - 7 796
AK152098.1 - 7 796

View Full Product Details