Gene Transcripts

Gene Symbol
Hoxc11
Entrez Gene ID
109663
Gene Name
homeobox C11
Gene Aliases
Hox-3.7
Chromosome Location
Chr. 15: 102954526 - 102956701 on Build GRCm38
UniGene ID
Mm.37692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024842.1 NP_001020013.1 1 20
GenBank mRNA BC139233.1 - 1 66
BC139269.1 - 1 66 8862291

View Full Product Details