Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009605.4 | NP_033735.3 | 3 | 756 | |
GenBank mRNA | AK003138.1 | - | 3 | 674 | |
AK041214.1 | - | 3 | 706 | ||
AK134112.1 | - | 3 | 756 | ||
AX767955.1 | - | 2 | 658 | ||
AX767957.1 | - | 3 | 687 | ||
AY749429.1 | - | 2 | 642 | ||
AY754346.1 | - | 2 | 642 | ||
BC028770.1 | - | 3 | 671 | 1329799 | |
U37222.1 | - | 3 | 687 | ||
U49915.1 | - | 2 | 658 |