Gene Transcripts

Gene Symbol
Cdc25a
Entrez Gene ID
12530
Gene Name
cell division cycle 25A
Gene Aliases
D9Ertd393e
Chromosome Location
Chr. 9: 109875337 - 109893895 on Build GRCm38
UniGene ID
Mm.307103
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007658.3 NP_031684.3 12 1512
GenBank mRNA AK036499.1 - 2 964
AK077609.1 - 12 1130
AK077991.1 - 12 1514
AK142334.1 - 11 1787

View Full Product Details