Gene Transcripts

Gene Symbol
Cdkn1b
Entrez Gene ID
12576
Gene Name
cyclin-dependent kinase inhibitor 1B
Gene Aliases
AA408329, AI843786, Kip1, p27, p27Kip1
Chromosome Location
Chr. 6: 134920401 - 134925525 on Build GRCm38
UniGene ID
Mm.2958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009875.4 NP_034005.2 1 708
GenBank mRNA AK046676.1 - 1 443
AK047669.1 - 1 709
AK050240.1 - 1 695
AK158411.1 - 1 688
AK162146.1 - 1 428
BC014296.1 - 1 691 4193493
CT010382.1 - 1 189
U09968.1 - 1 189
U10440.1 - 1 406

View Full Product Details