Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007807.5 | NP_031833.3 | 7 | 782 | |
GenBank mRNA | AK033570.1 | - | 7 | 726 | |
AK053920.1 | - | 7 | 726 | ||
AK152257.1 | - | 7 | 753 | ||
AK153154.1 | - | 7 | 753 | ||
AK153259.1 | - | 7 | 753 | ||
AK154126.1 | - | 7 | 782 | ||
AK160031.1 | - | 2 | 184 | ||
AK171526.1 | - | 7 | 768 | ||
BC042838.1 | - | 7 | 749 | ||
BC071229.1 | - | 7 | 737 | 4979804 | |
FJ168469.1 | - | 7 | 771 | ||
U43384.1 | - | 7 | 708 |