Gene Transcripts

Gene Symbol
Dgcr6
Entrez Gene ID
13353
Gene Name
DiGeorge syndrome critical region gene 6
Gene Aliases
Chromosome Location
Chr. 16: 18052860 - 18071632 on Build GRCm38
UniGene ID
Mm.27155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289813.1 NP_001276742.1 5 679
NM_001289814.1 NP_001276743.1 5 670
NM_010047.4 NP_034177.1 6 867
GenBank mRNA AF021031.1 - 5 571
AK019346.1 - 6 870
AK160342.1 - 6 857
AK185741.1 - 2 89
AK189125.1 - 2 89
BC048503.1 - 5 522 6770478

View Full Product Details