Gene Transcripts
- Gene Symbol
- Gys1
- Entrez Gene ID
- 14936
- Gene Name
- glycogen synthase 1, muscle
- Gene Aliases
- Gys3, MGS
- Chromosome Location
- Chr. 7: 45434839 - 45456617 on Build GRCm38
- UniGene ID
- Mm.275654
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030678.3 |
NP_109603.2
|
13, 14 |
1854 |
|
GenBank mRNA |
AK050813.1 |
- |
11, 12 |
1287 |
|
AK085476.1 |
- |
9, 10 |
882 |
|
AK169344.1 |
- |
13, 14 |
1850 |
|
AK170404.1 |
- |
13, 14 |
1856 |
|
AK170585.1 |
- |
13, 14 |
1774 |
|
AK171148.1 |
- |
13, 14 |
1809 |
|
AK171426.1 |
- |
13, 14 |
1833 |
|
BC019389.1 |
- |
13, 14 |
1647 |
3663506 |
BC131687.1 |
- |
13, 14 |
1701 |
40111722 |
BC131688.1 |
- |
13, 14 |
1701 |
40111723 |
BC152550.1 |
- |
13, 14 |
1701 |
40111724 |
U53218.1 |
- |
13, 14 |
1692 |
|
X94616.1 |
- |
13, 14 |
1701 |
|
View Full Product Details