Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207105.3 | NP_996988.2 | 2 | 204 | |
GenBank mRNA | AK077436.1 | - | 2 | 192 | |
AK170370.1 | - | 2 | 186 | ||
AY303784.1 | - | 1 | 37 | ||
AY303785.1 | - | 1 | 28 | ||
BY205244.1 | - | 2 | 223 |