Gene Transcripts

Gene Symbol
Hdgf
Entrez Gene ID
15191
Gene Name
hepatoma-derived growth factor
Gene Aliases
AI118077, D3Ertd299e
Chromosome Location
Chr. 3: 87906321 - 87916132 on Build GRCm38
UniGene ID
Mm.292208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008231.4 NP_032257.3 4 712
GenBank mRNA AF251787.1 - 4 625
AK005359.1 - 4 491
AK017863.1 - 4 666
AK029475.1 - 4 456
AK076021.1 - 4 500
BC005713.1 - 4 622 2609261
BC021654.1 - 4 640 4163897
D63707.1 - 4 466

View Full Product Details