Gene Transcripts

Gene Symbol
Ier3
Entrez Gene ID
15937
Gene Name
immediate early response 3
Gene Aliases
AI663993, IEX-1, cI-3, gly96
Chromosome Location
Chr. 17: 35821713 - 35822911 on Build GRCm38
UniGene ID
Mm.25613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133662.2 NP_598423.1 2 475
GenBank mRNA AK003466.1 - 2 501
AK051003.1 - 2 505
AK170477.1 - 2 505
BC006950.1 - 2 475 2811734

View Full Product Details