Gene Transcripts
- Gene Symbol
- Klrc1
- Entrez Gene ID
- 16641
- Gene Name
- killer cell lectin-like receptor subfamily C, member 1
- Gene Aliases
- CD159a, NKG2A, NKG2B
- Chromosome Location
- Chr. 6: 129666015 - 129682852 on Build GRCm38
- UniGene ID
- Mm.56899
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010652.2 |
NP_034782.1
|
2, 3 |
355 |
|
GenBank mRNA |
AF109784.1 |
- |
2, 3 |
292 |
|
View Full Product Details