Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_130450.2 | NP_569717.1 | 4 | 476 | |
GenBank mRNA | AB072039.1 | - | 4 | 423 | |
AF480860.1 | - | 4 | 395 | ||
AK029029.1 | - | 4 | 478 | ||
AK165847.1 | - | 3 | 500 | ||
AK167373.1 | - | 4 | 442 | ||
AY053453.1 | - | 4 | 457 | ||
BC040097.1 | - | 4 | 418 | ||
BC051041.1 | - | 4 | 473 | 4012676 | |
BC098492.1 | - | 3 | 473 | 30091954 | |
BC100576.1 | - | 4 | 418 | 30097278 |