Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133666.3 | NP_598427.1 | 7 | 1089 | |
GenBank mRNA | AK005021.1 | - | 7 | 1037 | |
AK075692.1 | - | 7 | 1034 | ||
AK151455.1 | - | 7 | 1000 | ||
AK178320.1 | - | 3 | 277 | ||
AK187850.1 | - | 3 | 277 | ||
AK188080.1 | - | 2 | 172 | ||
AK188929.1 | - | 2 | 159 | ||
AK205680.1 | - | 2 | 150 | ||
AK209435.1 | - | 3 | 277 | ||
AK210493.1 | - | 3 | 277 | ||
AK218408.1 | - | 3 | 276 | ||
BC014818.1 | - | 7 | 1059 | 4456931 | |
BC041682.1 | - | 7 | 992 | 4221001 |