Gene Transcripts

Gene Symbol
Nr4a2
Entrez Gene ID
18227
Gene Name
nuclear receptor subfamily 4, group A, member 2
Gene Aliases
HZF-3, NOT, Nurr1, RNR-1, TINOR, TINUR
Chromosome Location
Chr. 2: 57106830 - 57124003 on Build GRCm38
UniGene ID
Mm.3507
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139509.1 NP_001132981.1 4, 5 1299
NM_013613.2 NP_038641.1 4, 5 1445
GenBank mRNA AB014889.1 - 4, 5 1038
AK087495.1 - 5, 6 1323
AK158607.1 - 4, 5 1446
BC137715.1 - 4, 5 1757 8860734
BC144854.1 - 4, 5 1973
S53744.1 - 4, 5 1336

View Full Product Details