Gene Transcripts

Gene Symbol
Pfdn2
Entrez Gene ID
18637
Gene Name
prefoldin 2
Gene Aliases
ESTM27, W48336
Chromosome Location
Chr. 1: 171345699 - 171358170 on Build GRCm38
UniGene ID
Mm.10756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011070.3 NP_035200.2 3 238
GenBank mRNA BC026839.1 - 3 225
BC049606.1 - 3 238 6535749
Y17393.1 - 3 228

View Full Product Details