Gene Transcripts
- Gene Symbol
- Pgk1
- Entrez Gene ID
- 18655
- Gene Name
- phosphoglycerate kinase 1
- Gene Aliases
- Pgk-1
- Chromosome Location
- Chr. X: 106187100 - 106203699 on Build GRCm38
- UniGene ID
- Mm.316355
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008828.3 |
NP_032854.2
|
6, 7 |
785 |
|
GenBank mRNA |
AK133877.1 |
- |
6, 7 |
707 |
|
AK145846.1 |
- |
6, 7 |
721 |
|
AK164440.1 |
- |
6, 7 |
707 |
|
AK167441.1 |
- |
6, 7 |
679 |
|
AK167459.1 |
- |
6, 7 |
679 |
|
AK167710.1 |
- |
6, 7 |
678 |
|
BC023819.1 |
- |
6, 7 |
691 |
|
BC032167.1 |
- |
6, 7 |
691 |
|
BC083355.1 |
- |
6, 7 |
677 |
6828087 |
BC108372.1 |
- |
6, 7 |
689 |
5060103 |
M15668.1 |
- |
6, 7 |
656 |
|
View Full Product Details