Gene Transcripts

Gene Symbol
Plaa
Entrez Gene ID
18786
Gene Name
phospholipase A2, activating protein
Gene Aliases
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3
Chromosome Location
Chr. 4: 94565139 - 94603247 on Build GRCm38
UniGene ID
Mm.490101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172695.2 NP_766283.2 14 2136
GenBank mRNA AK019070.1 - 14 2143
AK075934.1 - 14 2068
AK085027.1 - 4 1583
BC139355.1 - 14 2135 8862377
BC139356.1 - 14 2135 8862378
BC139773.1 - 14 2046 40111789
DV646766.1 - 6 622

View Full Product Details