Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011171.2 | NP_035301.2 | 2 | 447 | |
GenBank mRNA | AK136541.1 | - | 2 | 307 | |
AK143719.1 | - | 2 | 549 | ||
AK167302.1 | - | 2 | 387 | ||
BC028755.1 | - | 2 | 320 | 1054063 | |
CT010175.1 | - | 2 | 266 | ||
L39017.1 | - | 2 | 321 | ||
X63748.2 | - | 2 | 353 |