Gene Transcripts

Gene Symbol
Ptpru
Entrez Gene ID
19273
Gene Name
protein tyrosine phosphatase, receptor type, U
Gene Aliases
Ftp-1, PCP-2, PTP, PTP-lambda, PTPlambda, Pcp2, Ptpf, Ptprl, R-PTP-U, R-PTP-psi, RPTPlambda
Chromosome Location
Chr. 4: 131768457 - 131838316 on Build GRCm38
UniGene ID
Mm.4860
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083119.2 NP_001076588.1 2 259
NM_011214.2 NP_035344.2 2 259
GenBank mRNA AK052720.1 - 2 234
AK148515.1 - 2 256
BC048694.1 - 2 215
BC080736.1 - 2 214
D88187.1 - 2 250

View Full Product Details