Gene Transcripts

Gene Symbol
Pirt
Entrez Gene ID
193003
Gene Name
phosphoinositide-interacting regulator of transient receptor potential channels
Gene Aliases
A530088H08Rik
Chromosome Location
Chr. 11: 66911910 - 66929877 on Build GRCm38
UniGene ID
Mm.440256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178656.3 NP_848771.1 2 409
GenBank mRNA AK041182.1 - 2 410
AK051158.1 - 2 355
AK051285.1 - 2 419
AK051645.1 - 2 410
AK084056.1 - 2 374
BC147296.1 - 2 307 9056340
BC147297.1 - 2 307 9056338
EU447173.1 - 2 400

View Full Product Details