Gene Transcripts

Gene Symbol
Slfnl1
Entrez Gene ID
194219
Gene Name
schlafen like 1
Gene Aliases
4933406A14Rik
Chromosome Location
Chr. 4: 120531981 - 120536661 on Build GRCm38
UniGene ID
Mm.29788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177570.3 NP_808238.1 2 523
GenBank mRNA AK077106.1 - 2 524
BC117545.1 - 1 274 40105640
BC118052.1 - 1 274 40105639

View Full Product Details