Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290122.1 | NP_001277051.1 | 3 | 1082 | |
NM_001290123.1 | NP_001277052.1 | 3 | 1071 | ||
NM_009032.3 | NP_033058.2 | 3 | 1118 | ||
GenBank mRNA | AK049659.1 | - | 3 | 1079 | |
BC130256.1 | - | 3 | 958 | 40142593 | |
BC144949.1 | - | 3 | 922 | 9053497 | |
U89506.1 | - | 4 | 957 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290127.1 | NP_001277056.1 | 3 | 1351 | |
NM_001290128.1 | NP_001277057.1 | 2 | 928 | ||
GenBank mRNA | EU287940.1 | - | 2 | 802 | |
EU287941.1 | - | 3 | 1225 |