Gene Transcripts
- Gene Symbol
- Six5
- Entrez Gene ID
- 20475
- Gene Name
- sine oculis-related homeobox 5
- Gene Aliases
- Dmahp, MDMAHP, TrexBF
- Chromosome Location
- Chr. 7: 19094544 - 19098549 on Build GRCm38
- UniGene ID
- Mm.3410
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011383.1 |
NP_035513.1
|
2 |
1599 |
|
GenBank mRNA |
D83146.1 |
- |
2 |
1350 |
|
View Full Product Details